TOC gas generators
TOC gas generators produce up to 1500 ml/min of carrier/combustion gas from in house compressed air supply. They are recommended for NPOC (Non-purgeable organic carbon). They have a CO₂ content of less than 1,0 ppm and a pressure dew point of −70 °C.
- Few moving parts with minimum maintenance requirements
- Flexibility to position generator where required, wall- mountable for small labs
- Completely silent in operation
- Maximum inlet pressure of 6,9 to 8,2 bar
- Catalog No:
- PEAK40-1040
- Dimensions:
- 430×410×620 mm
- Weight:
- 64 kg
Frequently Bought Together