HPLC columns, MetaSil AQ C18
Supplier: VARIAN
Certificates
About this item
The MetaSil AQ materials are based on silica that is highly coated with hydrophobic alkyl chains interspersed with hydrophilic short-chain endcapping groups. These columns remain fully extended in aqueous phases, allowing increased interaction between samples and the bonded phase. These columns remain fully extended in aqueous phases, allowing increased interaction between samples and the bonded phase.
Frequently Bought Together