Biocide, Klercide™ low residue quat
Klercide Low Residue Quat Blended with WFI version is a solution with water for injection (WFI) and the pack is triple bagged. Low endotoxin levels are guaranteed.
- Sterile cleanroom biocide for all types of hard surfaces
- Effective at low concentrations and non corrosive
- Fully compatible and ideal for a rotational regime
- SteriShield Delivery System™ ensures 3 months in-use shelf life
Suitable for ISO Class 5; Grade A and B (microbial count) areas: cleanroom processed, sterile filled and double bagged. Solution of quaternary ammonium compound and water, with broad spectrum activity. Filtered to 0,2 µm.
Bactericidal EN 1276; Fungicidal EN 1650; Bactericidal and Fungicidal EN 13697.
: Bactericidal EN 1276; Fungicidal EN 1650; Bactericidal and Fungicidal EN 13697.
: Use biocides safely. Always read the label and product information before use
Frequently Bought Together