Pipette tips, Ovation®
Supplier: VISTALAB TECHNOLOGIES
New Product
Certificates
About this item
Ideal for use with Ovation and MLA pipettes.
- Made of 100% pure virgin PP
- Provide a perfect fit
- Deliver reliable performance
- Autoclavable
Frequently Bought Together