Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Racks
Tube Racks
Test tube rack
Test tube rack
212-6211
:
WAGNER MUNZ
Test tube rack
212-6211
:
WAGNER MUNZ
For tube Ø:
18 mm
No. of Holes:
12
Colour:
Blue
Environmentally Preferable:
Pk:
1
PP, blue.
Excellent chemical resistance
Excellent temperature resistance