1,3-Dimethylimidazolium dimethylphosphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 1,3-Dimethylimidazolium Dimethyl Phosphate
CAS Number:654058-04-5
:
Formula:C₇H₁₅N₂O₄P
MDL Number:MFCD08275369
Molecular Weight:222.18 g/mol
:M-Clarity™ quality level = MQ100
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together