1H-Pyrazole-3-boronic acid hydrate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:376584-63-3
:
Formula:C₃H₅BN₂O₂·H₂O
MDL Number:MFCD11044442
Molecular Weight:111.90 g/mol
Melting Pt:85…88 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together