Sumps for IBC station with platform
: asecos
Polyethylene sumppallet with 1000 litres capacity for the storage of IBC.
- Complete corrosion protection
- Optionally with integrated dispensing area
- Integrated polyethylene storage platform acts as a safety measure
Compact and robust design. Safe and approved for the safe storage of water pollutants.
Frequently Bought Together