Order Entry
Ireland
ContactUsLinkComponent
Potassium bis(trimethylsilyl)amide 15% (w/w) 0.7 M in toluene, AcroSeal®
Potassium bis(trimethylsilyl)amide 15% (w/w) 0.7 M in toluene, AcroSeal®
Certificates
undefined
Potassium bis(trimethylsilyl)amide 15% (w/w) 0.7 M in toluene, AcroSeal®
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear to slightly hazy colorless to amber Solution
  • Titration with HClO4:
    0.6 to 0.9 M
  • Melting point:
    83 to 86 °C
  • GC:
    ≥95 %

Specification Test Results

Frequently Bought Together