About this item
These kits provide the user with printer and a selection of labels to accurately identify samples in the laboratory. Both the BMP™51 and BMP™53 are portable, single colour, thermal printers with 300 dpi print head and with very similar specifications, except the BMP™51 has stand alone capability and the BMP™53 does not. The top-loading label cartridge contains both the printer ribbon and labels and can be exchanged in seconds.
- USB, Bluetooth and optional Ethernet, WiFi connectivity
- Print speed 25,4 mm/second
- BMP™51 Laboratory identification kit has qwerty keyboard for on the spot sample identification
Laboratory identification kit: BMP™51 or BMP™53 Printer are supplied with hard case, USB cable, rechargeable Li-on battery and AC adaptor with power cord; vial and tube labels (M-122-461), microplate labels (M-131-499); general id label (MC-1500-595-WT-BK). Optional Brady LabelMark™ 5 labelling software is available separately .