4-Methoxy-N-methylaniline 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: N-Methyl-p-anisidine
CAS Number:5961-59-1
:
Formula:C₈H₁₁NO
MDL Number:MFCD00008399
Boiling Pt:135…136 °C (19 mmHg)
Molecular Weight:137.18 g/mol
Melting Pt:33…36 °C
Flash Pt:110 °C
EINECS:227-735-1
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to grey-brown Low melting crystalline mass
- Infrared spectrum:Conforms
- Melting point:33 to 36 °C
- GC:≥97.5 %
- HPLC:≥98.5 %
Specification Test Results
Frequently Bought Together