Order Entry
Ireland
ContactUsLinkComponent
Hafnium disilicide
Hafnium disilicide
Catalog # 12564.09
CAS Number:  
Hafnium disilicide
Catalog # 12564.09
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Hafnium silicide
 :
 :234.66 g/mol
Storage Temperature:Ambient
EINECS:235-640-1
CAS Number:12401-56-8
Formula:HfSi₂
MDL Number:MFCD00167026