Caps with dropper inserts for bottles
Supplier: VITLAB
Certificates
About this item
Made from HDPE.
- Extra long, fine dropping tip for accurate dispensing
- Good chemical resistance
- Good temperature resistance
For bottles with GL threads. Cap with dropper insert, complete with screw cap.
Frequently Bought Together