Organic reference standard, Ethylene oxide (Oxirane) 50,000 µg/ml in methanol, Supelco®
Catalog # 48838.
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass ampoule
- Pk:1 Ampoul
- CAS No.:75-21-8
- Description:Ethylene oxide (Oxirane) 50,000 µg/ml in methanol
- Environmentally Preferable:
- Formula:C₂H₄O
- Description:Ethylene oxide (Oxirane)