2-Propanol ≥98%, TECHNICAL
: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
:67-63-0
:
:(CH₃)₂CHOH
:MFCD00011674
:82
:–89 °C
:0.786 g/cm³ (20 °C)
:60.1 g/mol
:12
:Ambient
:1219
:3 II
:13,05228
:200-661-7
:01-2119457558-25
:GHS02
:GHS07
- Environmentally Preferable:
- Assay:Min. 98 %
Frequently Bought Together