Anti-CD79a Mouse Recombinant Antibody [clone: rIGA/6986]
Catalog # ANTIA316134-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:B lymphocyte-specific MB1 protein
- Antigen symbol:CD79a
- Clonality:Recombinant
- Clone:rIGA/6986
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P11912
- Antigen synonyms:Ig-alpha|CD79A antigen|CD79a antigen (immunoglobulin-associated alpha)|CD79a|CD 79a|B-cell antigen receptor complex-associated protein alpha chain|CD79A_HUMAN|Ig alpha|CD79a molecule, immunoglobulin-associated alpha
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:44 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-100) of human CD79a protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rIGA/6986] antibody to CD79a for WB and IHC-P with samples derived from human.
- Validated applications: WB, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ ml, IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: CD79a
Clonality: Recombinant
Clone: rIGA/6986
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human