Pyridinium chloride 98%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Pyridine hydrochloride, Pyridinehydrochloride, Pyridine HCl
CAS Number:628-13-7
:
Formula:C₅H₆ClN
MDL Number:MFCD00012802
Boiling Pt:222.5
Molecular Weight:115.56 g/mol
Melting Pt:140…146 °C
EINECS:211-027-4
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to tan Crystals
- Infrared spectrum:Conforms
- Titration Argentometric:≥97.5 %
- Solubility:(5% in water) Clear colorless to
Specification Test Results
Frequently Bought Together