Hybridoma growth and expression CD media HYGM7
Supplier: BIOCONCEPT
New Product
Certificates
About this item
Hybridoma growth media (HYGM) 7 is a serum-free, protein free, chemically defined (CD) and fully defined media that can be used for the cultivation of various different hybridomas and the production of monoclonal antibodies.
- Chemically Defined (CD)
- Serum-free
- Protein free
- Ready to use
- High yielding
- Contsins therapeutic grade recombinant insulin
- Free of hydrolysates or peptones
- Free of any undefined components
Certifications: ISO 9001; cGMP.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together