Flat head mop wringer, MegaWringer™
Supplier: MICRONOVA
Certificates
About this item
A stainless steel flat head mop wringer with a unique design that leverages the mop handle to exert downward wringing pressure, doing away with the need to bend and press while cleaning.
- Down-press wringer, ideal for consistent wringing
- One hand action, easy to use with minimal pressure
- Guards against operator wrist and back strain
- Unit is compact, easily transported and stored, and weighs less than 5 pounds
- Electropolished stainless steel, with moulded polyketone components
- Pair with Slim T™ buckets for cart-free use
- Holds up to strong disinfectants
- Autoclavable
The Micronova MegaWringer™ flat head mop wringer features a simple, sturdy design that leave mops damp enough to effectively disinfect surfaces, while wringing out excess liquid. Designed to be used with Slim T™ buckets in BucketBinder™ systems or Slim T™ cart systems.
Specifications
- Catalog No:
- 216-2571
- 216-1491
- 129-0064
- 141-0107
- Environmentally Preferable:
- Sustainable Materials, Sustainable Packaging, Water Efficient, Energy Efficient