Syringaldazine
: 4-[[(3,5-dimethoxy-4-oxo-1-cyclohexa-2,5-dienylidene)methylhydrazo]methylidene]-2,6-dimethoxy-1-cyclohexa-2,5-dienone, 4-Hydroxy-3,5-dimethoxybenzaldehyde azine, 3,5-Dimethoxy-4-hydroxybenzalazine, 4,4'-(Hydrazine-1,2-diylidenebis(methanylylidene))bis(2,6-dimethoxyphenol)
:14414-32-5
:
:C₁₈H₂₀N₂O₆
:MFCD00008363
:360.36 g/mol
:209…210 °C
:GHS07
:Refrigerator
- Environmentally Preferable:
- Appearance:Yellow to green Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:209 to 213 °C
- HPLC:≥96.0 %
Frequently Bought Together