Order Entry
France
ContactUsLinkComponent
VTM - virus transport medium
VTM - virus transport medium
Catalog # LIOF26490
Supplier:  LIOFILCHEM
CAS Number:  
undefined
VTM - virus transport medium
Catalog # LIOF26490
Supplier:  LIOFILCHEM
CAS Number:  

Specifications

  • Pk:
    100
  • Description:
    1 pack × 100 Plastic tubes
  • Environmentally Preferable:
  • Storage conditions:
    10…25° C

Specifications

About this item

VTM is suitable for collection and preservation of clinical specimens containing viruses, including SARS-CoV-2 (COVID-19), Chlamydia spp., Mycoplasma spp. and Ureaplasma spp.

  • Storage at 10…25° C
  • Contains antimicrobial agents to minimize bacterial and fungal contamination, without affecting viruses, Chlamydia, Mycoplasma or Ureaplasma
  • A pH indicator (red phenol) indicates the medium integrity during the shelf life of the product
  • Shelf life: 1 year
  • The tube stands upright on the laboratory bench thanks to the skirted, flat-bottom, while the conical shape allows an appropriate centrifugation of the sample.
  • 16 x 100 mm skirted tube with plastic red cap, sterile

The transport medium can be used with swabs and aspirates and is available in a plastic, screw cap tube. The transported microorganism remains viable for 48 hours at room or refrigerated temperature. Every tube contains 3ml of medium.