Magnesium sulphate ≥95%, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: MgSO4, Magnesium sulphate anhydrous
CAS Number:7487-88-9
:
Formula:MgSO₄
MDL Number:MFCD00011110
Density:2.65 g/cm³ (20 °C)
Molecular Weight:120.37 g/mol
Melting Pt:1127 °C
Merck Index:12,05731
Storage Temperature:Ambient
EINECS:231-298-2
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 95 %
Specification Test Results
Related Information
Certificates
Search by Lot Number25162_MAGNESIUM SULPHATE_BSE_REV0
BSE/TSE Certificates
27571126.pdf 27914431.pdf spec_VWRC25162.361 spec_VWRC25162.361 spec_VWRC25162.465
Product specification
33L Plastic bucket UN Z (ZPACK13247037) 5L Plastic bucket UN Z (ZPACK13247005)
Packaging Information
VWR® for Drying and Filtration
Product Literature
Frequently Bought Together