Order Entry
France
ContactUsLinkComponent
 
Benzyl salicylate
Catalog # EHERC16903510
Supplier:  EHRENSTORFER
CAS Number:  
Benzyl salicylate
Catalog # EHERC16903510
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Benzyl 2-hydroxybenzoate
 :
 :228.25 g/mol
EINECS:204-262-9
CAS Number:118-58-1
Formula:C₁₄H₁₂O₃
MDL Number:MFCD00020034
Boiling Pt:208
Melting Pt:23.9
Density:1.1755 g/cm³ (20 °C)
Merck Index:13,01146