Order Entry
France
ContactUsLinkComponent
Anti-CFC1 Goat Polyclonal Antibody
Anti-CFC1 Goat Polyclonal Antibody
  PRSI45-432
undefined
Anti-CFC1 Goat Polyclonal Antibody
  PRSI45-432

 

  • Antigen symbol:
    CFC1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    Liquid
  • Concentration:
    500 ug/mL
  • Immunogen:
    Cripto-1 antibody was raised against a 16 amino acid synthetic peptide near the internal region of Cripto-1.
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Cripto-1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Cripto-1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CFC1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: