Specifications
- Antibody type:Primary
- Antigen name:Dynein Axonemal Intermediate Polypeptide 1 and 2
- Antigen symbol:DNAI1/2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:DNAI 1|ICS|DNAI1_HUMAN|MGC26204|DNAI 2|Axonemal dynein intermediate chain 2|PCD.|DNAI1|Dynein axonemal intermediate chain 1|CILD 1|Dynein intermediate chain 1 axonemal|Dynein intermediate chain 1|Dynein axonemal intermediate polypeptide 2|Axonemal dynein intermediate chain 1|CILD1|Dynein intermediate chain 2 axonemal|Dynein intermediate chain DNAI1|axonemal|Dynein axonemal intermediate polypeptide 1|Dynein intermediate chain 1 cytosolic|Dynein intermediate chain 2 cytosolic
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The inner- and outer-arm dyneins, which bridge between the doublet microtubules in axonemes, are the force-generating proteins responsible for the sliding movement in axonemes. The intermediate and light chains, thought to form the base of the dynein arm, help mediate attachment and may also participate in regulating dynein activity. This gene encodes an intermediate chain dynein, belonging to the large family of motor proteins. Mutations in this gene result in abnormal ciliary ultrastructure and function associated with primary ciliary dyskinesia (PCD) and Kartagener syndrome. [provided by RefSeq, Jul 2008].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DNAI1/2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat