Order Entry
France
ContactUsLinkComponent
Tissue DNA kit, genomic DNA isolation from tissue, E.Z.N.A.®
Tissue DNA kit, genomic DNA isolation from tissue, E.Z.N.A.®
Supplier:  OMEGA BIO-TEK
Certificates
undefined
Tissue DNA kit, genomic DNA isolation from tissue, E.Z.N.A.®
Supplier:  OMEGA BIO-TEK
Certificates

Specifications

  • Application:
  • Method / Format:
  • Refine target molecule:
  • Sample type:
  • Target molecule:
  • Environmentally Preferable:

Specifications

Related Information

Frequently Bought Together