Pyruvonitrile 90%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:631-57-2
:
Formula:C₃H₃NO
MDL Number:MFCD00001838
Boiling Pt:92…93 °C
Density:0.974
Molecular Weight:69.063 g/mol
Flash Pt:14 °C (57 °F)
EINECS:211-159-2
UN:3273
ADR:3 II
:GHS02
:GHS06
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together