Anti-4-HNE Monoclonal Antibody [clone: HNEJ-2]
Catalog # ABNOMAB6115
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:4-HNE
- Clonality:Monoclonal
- Clone:HNEJ-2
- Conjugation:Unconjugated
- Host:
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Lyophilized
- Storage buffer:50 mM TBS (1% BSA)
- Storage temperature:Store at 4 °C on dry atmosphere. After reconstitution with deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:4-HNE-modified conjugated with KLH.
- Size:20 μg
- Pk:20 µg
Specifications
About this item
Mouse monoclonal antibody raised against 4-HNE.
Recommended Dilutions: Immunohistochemistry: 25 ?g/ml; Western Blot: 15 ?g/ml
Type: Primary
Antigen: 4-HNE
Clonality: Monoclonal
Clone: HNEJ-2
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: