Anti-Glycogen Synthase Kinase 3 alpha, beta Mouse monoclonal antibody [clone: 5J105]
Catalog # USBIG8170-41A1
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Glycogen Synthase Kinase 3 alpha and beta
- Antigen symbol:GSK3A/GSK3B
- Clonality:Monoclonal
- Clone:5J105
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human,Xenopus laevis,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,01% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:10 Pieces
- Pk:10
Specifications
About this item
Recommended dilution: ELISA: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: Glycogen Synthase Kinase 3 alpha, beta
Clonality: monoclonal
Clone: 5J105
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Xenopus laevis