Anti-Melanoma, gp100 Mouse Monoclonal Antibody [clone: HMB45]
USBI138570-500
- Antibody type:Primary
- Antigen name:Melanoma gp100
- Clonality:Monoclonal
- Clone:HMB45
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Format:Supernatant
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µl
- Pk:500 µl
Recommended dilution: Immunofluorescence: 1:50-1:100; Western Blot: 1:100-1:200; Immunoprecipitation: 5-10?l/500 ?g; Immunochemistry: 1:100-1:200
Type: Primary
Antigen: Melanoma, gp100
Clonality: monoclonal
Clone: HMB45
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human