Order Entry
France
ContactUsLinkComponent
Anti-CTNA1 Rabbit Polyclonal Antibody
Anti-CTNA1 Rabbit Polyclonal Antibody
Catalog # ABGEAP6582A
Supplier:  Abgent
undefined
Anti-CTNA1 Rabbit Polyclonal Antibody
Catalog # ABGEAP6582A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Catenin Alpha 1
  • Antigen symbol:
    CTNNA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-terminal
  • Antigen synonyms:
    Alpha N-catenin|Alpha-catenin-related protein|CTNNA2|Catenin alpha2|Catenin alpha1|Renal carcinoma antigen NY-REN-13,Catenin-alpha1|alphaN-catenin|Catenin-alpha2|alphaE-catenin|Catenin alpha-1|CAPR|Cadherin-associated protein|Catenin alpha-2|Alpha E-catenin
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: CTNA1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse