Anti-GRB2 Rabbit Polyclonal Antibody
Catalog # ABGEAP6283C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Growth factor receptor-bound protein 2
- Antigen symbol:GRB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Antigen synonyms:Adapter protein GRB2|SH2/SH3 adapter GRB2|Protein Ash|ASH
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: GRB2 (growth factor receptor-bound protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human