Anti-AFF2 Rabbit Polyclonal Antibody
Catalog # AVIVARP38099_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:AF4/FMR2 family, member 2
- Antigen symbol:AFF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:2334
- Antigen synonyms:FRAXE|FMR2|MRX2|OX19|FMR2P
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: AFF2 (AF4/FMR2 family, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: