Order Entry
France
ContactUsLinkComponent
Antiseptic soap, Dermanios Scrub CG
  DMDA2082522
Antiseptic soap, Dermanios Scrub CG
  DMDA2082522
  • Description:
    Antiseptic soap, Dermanios Scrub CG, bottle with screw down pump, 500 ml
  • Seq:
    02
  • Environmentally Preferable:
  • Pk:
    12
Antiseptic soap for hygienic handwash and general body cleansing.
  • Ready to use product
  • Bactericidal, yeasticidal, virusidal (Herpes virus , BVDV ( model virus HCV ), RSV)
 : EN 1040 , pr EN 13727, EN 1499, EN 1275, EN 1650
 : Use biocides safely. Always read the label and product information before use.