Aspirator bottles, glass, with stopper, PYREX®
Supplier: SciLabware
Certificates
About this item
Made of PYREX®, borosilicate glass.
- Ground side socket (size 29/32) to accept stopcock assemblies
- Ground neck
- Supplied with glass stopper
Bottle with grounded side-socket.
Certifications: Complies with ISO 4796