Screw caps for laboratory bottles
Screw cap, made of PP, for media-lab bottles
- Plug seal cap ideal for media preparation work
- Autoclavable up to 140°C
- Available in choice of 4 colours for easy identification of samples
Screw cap, made of PP, for media-lab bottles