ContactUsLinkComponent
VWR®, Weighing Table, Granite, for Micro and Analytical Balances
VWR®, Weighing Table, Granite, for Micro and Analytical Balances
  611-2549
undefined
VWR®, Weighing Table, Granite, for Micro and Analytical Balances
  611-2549
  • Description:
    Balance table, granite, with vibration dampers and adjustable feet
  • W×D×H:
    900×600×760 mm
  • Environmentally Preferable:
  • Pk:
    1
  • Weight:
    267 kg
High performance micro balances and analytical balances should be placed in an environment where interfering vibration is at a minimum, to give fast reading stabilisation. The VWR® weighing table features a sturdy, stable frame.
  • Granite weighing surface
  • Adjustable feet
  • Meets highest anti-vibration standards, with four vibration dampers located under the work surface
  • Facilitates fast stabilisation times
 : Installation not included.