Petri dishes, square, Sterilin®
Supplier: Sterilin
Certificates
About this item
Non compartmentalised dish is ideal for antibiotic sensitivity testing when a large surface area and very flat base is required.
- Ideal for antibiotic sensitivity testing when a large surface area and very flat base is required
- Ideal for small volume liquid media work or for sample storage
- Lid features selectable venting or non-venting positions where the lid turns through 90° to allow for venting or non-venting of the dish
Compartmentalised dish is ideal for small volume liquid media work or for sample storage. The 25 compartments each have a surface area of 1,8 cm² and a capacity of 5 ml. The lid features selectable venting or non venting positions.
Dimensions shown are for outer Ø base and outer height (mm).
Conduct various applications with Thermo Scientific™ Sterilin™ 100 mm square Petri dishes, available in both compartmentalised and non-compartmentalised formats. Aseptically manufactured under clean room conditions (class 7 ISO14644) to exclude microbiological contamination. Not available in America.