3-Methyl-1-cyclohexene 90%, tech.
ACRO304310250
:
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
- Environmentally Preferable:
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4425 to 1.4445 (20°C, 589 nm)
- GC:≥88 %
: (±)-3-Methylcyclohexene
:
:96.17 g/mol
:104
:–3 °C
:209-718-0
:1993
:GHS02
:Ambient
:591-48-0
:C₇H₁₂
:MFCD00001571
:–120.4 °C
:0.8 g/cm³
:3 II