Glove bags
Supplier: abcr
Certificates
About this item
These durable PE glove bags effectively eliminate oxygen and moisture during equipment manipulation operations.
- To be used like a glove box; gloves are integral
- Equipment is placed in the bag through an entry sleeve, the bag is sealed, connected to the gas port and inflated
- Economical, since reusable