Anti-POLR3K Rabbit Polyclonal Antibody
PRSI22-246
New Product
- Antibody type:Primary
- Antigen name:RNA polymerase III subunit K
- Antigen symbol:POLR3K
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:51728
- Antigen synonyms:RNA polymerase III subunit CII|RPC12.5|C11-RNP3|polymerase (RNA) III subunit K|RNA polymerase III subunit C1RNA polymerase III subunit C11|DNA-directed RNA polymerases III 12.5 kDa polypeptide|12.3 kDa|C11|My01RPC1RPC11|RNA polymerase III 12.5 kDa subunit|polymerase (RNA) III (DNA directed) polypeptide K|DNA-directed RNA polymerase III subunit RPC1DNA-directed RNA polymerase III subunit K
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-108 of human POLR3K (NP_057394.3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: POLR3K
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human