6-Methylnicotinic acid 99%
: 6-Methyl-3-pyridinecarboxylic acid, 6-Methylpyridine-3-carboxylic acid
:3222-47-7
:
:C₇H₇NO₂
:MFCD00006341
:137.14 g/mol
:209…213 °C
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:White to light yellow to beige Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:209 to 213 °C
- Titration with NaOH:≥98.5 %