Tris(dimethylamino)phosphine ≥97%
: Tris(dimethylamino)phosphine, Hexamethyl phosphorous triamide, Hexamethyltriaminophosphine, HMPT
:1608-26-0
:
:C₆H₁₈N₃P
:MFCD00008301
:162…164 °C
:–44 °C
:0.898
:163.2 g/mol
:37 °C (98 °F)
:Ambient
:216-534-4
:3 III
:GHS08
:GHS02
- Environmentally Preferable: