Donepezil Quaternary Salt, Pharmaceutical Analytical Impurities
Catalog # 1A00250.
Supplier: USP
CAS Number:
Specifications
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:844694-85-5
- Description:Donepezil Quaternary Salt
- Environmentally Preferable:
- Synonym(s):Donepezilbenzyl Bromide) (1,1-dibenzyl-4-((5,6-dimethoxy-1-oxo-2,3-dihydro-1H-inden-2-yl)methyl)piperidin-1-ium, bromide; 1,1-dibenzyl-4-[(5,6-dimethoxy-1-oxoindan-2-yl)methyl]piperidinium, bromide
- Description:Donepezil Quaternary Salt
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.