2-Bromo-5-fluorophenylboronic acid
Catalog # APOSPC8649-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: (2-Bromo-5-fluorophenyl)boronic acid
:
:218.82 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:959996-48-6
Formula:C₆H₅BBrFO₂
MDL Number:MFCD08690232