Ethyl 1-phenyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Ethyl 1-phenyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate
CAS Number:741717-63-5
:
Formula:C₁₃H₁₁F₃N₂O₂
MDL Number:MFCD10568308
Molecular Weight:284.2339 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: