Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4'-Fluoro-3'-(trifluoromethyl)acetophenone
4'-Fluoro-3'-(trifluoromethyl)acetophenone 97%
4'-Fluoro-3'-(trifluoromethyl)acetophenone 97%
:
Apollo Scientific
4'-Fluoro-3'-(trifluoromethyl)acetophenone 97%
:
Apollo Scientific
:
1-(4-Fluoro-3-(trifluoromethyl)phenyl)ethanone
:
208173-24-4
:
:
C₉H₆F₄O
:
MFCD00061252
:
1.36 g/cm³ (25 °C)
:
206.14 g/mol
:
74 °C
:
GHS07
:
Ambient
Environmentally Preferable: