Imidazo[1,5-a]pyridine-1-carbaldehyde 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:56671-67-1
:
Formula:C₈H₆N₂O
MDL Number:MFCD06739249
Molecular Weight:146.150 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: