1-(3-Hydroxypropyl)piperazine 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 1-Piperazinepropanol, 3-Piperazin-1-ylpropanol, 3-(Piperazin-1-yl)propan-1-ol
CAS Number:5317-32-8
:
Formula:C₇H₁₆N₂O
MDL Number:MFCD00023132
Boiling Pt:128…132 °C/5.25mmHg
Molecular Weight:144.22 g/mol
Melting Pt:49…53 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: