2-Chloro-3-nitrophenol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:603-84-9
:
Formula:C₆H₄ClNO₃
MDL Number:MFCD11035882
Molecular Weight:173.5500 g/mol
Melting Pt:120 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: